Learn More
Med Vet International #4 3/8in Curved Heavy Duty Suture Needles, J0110PM4, 12/Pack
Supplier:  Med Vet International JOR110PM4
Encompass_Preferred
3/8in-curved. Stainless steel. Cutting edge. Length refers to actual length of needle.
Catalog No. 50-005-83803