Learn More
Medchemexpress LLC 4-(3-Chloro-4-(3-(cyclopropyl-2,2,3,3-d4)ureido)phenoxy)-7-methoxyquinoline-6-carboxamide-15N | 98.0% | 431.87 | 50 MG

Supplier: Medchemexpress LLC HY10981S21MG
Lenvatinib-15N,d4 is a chemical compound supplied as a white to off-white solid. This product has been tested and complies with the given specifications, showing consistency with its structure as confirmed by 1H NMR Spectrum and LCMS analysis.
- Molecular formula: C₂₁H₁₅D₄ClN₃¹⁵NO₄
- Molecular weight: 431.87
- Appearance: White to off-white solid
- Purity (LCMS): 98.01%
- Consistent with structure by ¹H NMR spectrum and LCMS
- Storage recommendations: -20°C for 3 years, 4°C for 2 years (powder); -80°C for 6 months, -20°C for 1 month (in solvent)
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.