Learn More
Medchemexpress LLC 4-(3-chloro-4-fluoroanilino)-6-(pyridin-3-ylmethylamino)-1,7-naphthyridine-3-carbonitrile | 871307-18-5 | 404.83 g/mol | 10mg
SDP
Supplier:  Medchemexpress LLC HY1235810MG
Encompass_Preferred
Tpl2 Kinase Inhibitor 1 is a 3-pyridylmethylamino analog and is a selective Tpl2 inhibitor (IC50 50 nM) Tpl2 consists of COT kinase and MAP3K8 Tpl2 Kinase Inhibitor 1 plays an important role in the regulation of the inflammatory response and the progression of some cancers[1][2]
Catalog No. 50-002-37686