Learn More
Medchemexpress LLC 4-(3-chloro-4-fluoroanilino)-6-(pyridin-3-ylmethylamino)-1,7-naphthyridine-3-carbonitrile | 871307-18-5 | MFCD12405303 | 99.4% | 404.83 g/mol | C21H14ClFN6 | 10MM 1ML

Supplier: Medchemexpress LLC HY1235810MM 1ML
Tpl2 Kinase Inhibitor 1 is a small-molecule research reagent that selectively inhibits Tpl2 (MAP3K8) with biochemical potency (IC50 = 50 nM) and demonstrated cellular activity in monocytes and whole blood. It is supplied as a solid and as a ready-to-use 10 mM solution in DMSO for studies of MAP3K-dependent signaling and inflammatory responses.
- Selective Tpl2 inhibition with IC50 ≈ 50 nM.
- Inhibits LPS-induced TNF-alpha production in human monocytes (IC50 ~0.7-1.1 μM) and whole blood (IC50 8.5 μM).
- Chemical formula C21H14ClFN6; molecular weight 404.83 g/mol.
- High purity (~99.4%).
- Available as solid and as 10 mM solution in DMSO (1 mL).
- Storage: powder at -20 °C; in solvent at -80 °C (6 months) or -20 °C (1 month).
- Intended for use as a research tool to probe inflammatory and cancer-related signaling pathways.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.