Learn More
Medchemexpress LLC 4-(3-fluoro-5-trifluoromethylpyridin-2-yl)piperazin-1-yl methanone | 845614-11-1 | MFCD18251496 | 543.46 g/mol | C21H20F7N3O4S | 10 MG

Supplier: Medchemexpress LLC HY1080910MG
Bitopertin is a research reagent that acts as a potent, selective inhibitor of the glycine transporter 1 (GlyT1). It reduces glycine reuptake at human GlyT1 with a reported IC50 of 25 nM and is supplied as a solid powder intended for research and analytical use.
- Potent, selective GlyT1 inhibitor with reported IC50 of 25 nM.
- Noncompetitive mechanism of action.
- Suitable for in vitro research and analytical applications.
- Provided as a solid powder with recommended storage guidance.
- Molecular formula C21H20F7N3O4S and molecular weight 543.46 g/mol.
- CAS number 845614-11-1 for unambiguous identification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.