Learn More
Medchemexpress LLC 4(3H)-quinazolinone | 491-36-1 | MFCD00511302 | 99.9% | 146.15 g·mol⁻¹ | C8H6N2O | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01880010G
Encompass_Preferred
4(3H)-Quinazolinone is a fused nitrogen heterocyclic compound used as a chemical building block in research and synthesis. Derivatives of this scaffold have shown antimicrobial and antitumor activity. The material is a white to off-white solid with formula C8H6N2O and molecular weight 146.15 g·mol⁻¹, supplied at high purity for laboratory applications.
  • High purity (≈99.9%) suitable for synthesis and analytical work.
  • Used as a building block for antimicrobial and antitumor quinazolinone derivatives.
  • Solid, white to off-white appearance for easy handling.
  • Available in multiple pack sizes including 1 g to 100 g and bulk options.
  • Includes standard structural identifiers (formula, SMILES, molecular weight).
Catalog No. 50-003-00039