Learn More
Medchemexpress LLC 4-[4-(1H-Imidazol-1-yl)phenyl]-N-2-propyn-1-yl-2-pyrimidinamine | 2234272-10-5 | 98.1% | C16H13N5 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13053010MG
Encompass_Preferred
AP-C5 is a selective small-molecule inhibitor of cGMP-dependent protein kinase II (cGKII) used in cellular and biochemical research, including studies of diarrheal disease. It also contains an alkyne functional group that enables copper-catalyzed azide-alkyne cycloaddition (click chemistry) for probe conjugation.
  • Selective cGKII inhibition suitable for biochemical and cellular assays.
  • Click-chemistry compatible alkyne for facile probe conjugation.
  • High purity (>98%) for consistent experimental performance.
  • Solid, light yellow to light brown physical form for straightforward handling.
  • Stable when stored under recommended conditions for long-term use.
  • Good solubility in DMSO for preparation of screening solutions.
Catalog No. 50-002-58861