Learn More
Medchemexpress LLC 4-[[4-(2-methoxyethoxy)cyclohexyl]amino]-1-methyl-6-(1,3-thiazol-5-yl)quinolin-2-one | 1700637-55-3 | MFCD32062812 | 99.3% | 413.53 | C22H27N3O3S | 500MG
SDP
Supplier:  Medchemexpress LLC HY123999500MG
Encompass_Preferred
CD38 inhibitor 1 (compound 78c) is a potent small-molecule inhibitor of the CD38 enzyme intended for research use. It shows low-nanomolar inhibitory activity against human and mouse CD38 and is supplied as a high-purity solid for biochemical and cellular studies.
  • Potent inhibition of CD38 with low-nanomolar IC50 values.
  • Active against both human and mouse CD38.
  • High purity suitable for research applications (reported ≥98%).
  • Available in multiple package sizes, including a 500 mg option.
  • Useful for studies of NAD+ metabolism and CD38-related pathways.
Catalog No. 50-002-38383