Learn More
Medchemexpress LLC 4-[4-[(2-thiiranylmethyl)sulfonyl]phenoxy]-benzenemethanamine monohydrochloride | 1807453-83-3 | MFCD34469331 | 97.2% | 371.90 | C16H18ClNO3S2 | 1MG

Supplier: Medchemexpress LLC HY1243731MG
ND-336 is a small-molecule selective inhibitor of matrix metalloproteinases MMP-2, MMP-9, and MMP-14 used in biochemical and in vivo research; it exhibits low-nanomolar to sub-micromolar potency and has been reported to accelerate diabetic wound healing in mice by reducing inflammation and promoting angiogenesis and re-epithelialization. The compound is supplied as a white to off-white solid, with high purity and recommended cold storage for long-term stability.
- Selective inhibitor of MMP-2, MMP-9, and MMP-14.
- Potency: Ki = 85 nM (MMP-2), 150 nM (MMP-9), 120 nM (MMP-14).
- Shown to accelerate wound healing in diabetic mouse models.
- High purity (97.2%) and white to off-white solid appearance.
- Soluble in DMSO at 30 mg/mL (requires ultrasonic and warming).
- Storage: solid at -20°C; in solvent -80°C for 6 months, -20°C for 1 month.
- Suitable for biochemical assays and topical in vivo studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.