Learn More
Medchemexpress LLC 4-(4-(4-(3-Aminophenyl)-1H-1,2,3-triazol-1-yl)phenoxy)-N-methylpicolinamide | 99.3% | 386.41 | 5 MG

Supplier: Medchemexpress LLC HY1690495MG
Smurf1 ligand 1 is a PROTAC target protein ligand. It can be used for the synthesis of SMART1.
- Purity: 99.31%
- Molecular weight: 386.41
- Unit of measure: 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg, 500 mg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.