Learn More
Medchemexpress LLC 4-(4,4-difluoro-1-(2-fluorophenyl)cyclohexyl)phenol | 1421854-16-1 | 99.6% | 306.32 | C18H17F3O | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1101575MG
Encompass_Preferred
AC-186 is a selective, non-steroidal estrogen receptor β (ERβ) agonist used for preclinical research. It exhibits potent ERβ activity (EC50 ≈ 6 nM) with low ERα activity (EC50 ≈ 5,000 nM) and has shown gender-selective neuroprotective effects in male rat models of Parkinson's disease.
  • Selective ERβ agonist activity for receptor pharmacology.
  • Demonstrated neuroprotection in preclinical Parkinson's disease studies.
  • High purity for consistent experimental results (~99.6%).
  • Defined chemical properties and molecular weight for analytical work.
  • Available in research-scale packaging suitable for laboratory use.
Catalog No. 50-002-63558