Learn More
Medchemexpress LLC 4-(((4-(5-chloro-2-(((1r,4r)-4-((2-methoxyethyl)amino)cyclohexyl)amino)pyridin-4-yl)thiazol-2-yl)amino)methyl)... | 2247481-21-4 | 98.7% | 505.08 | C24H33ClN6O2S | 1mL
SDP
Supplier:  Medchemexpress LLC HYX015010MM/1ML
Encompass_Preferred
JSH-150 is a highly selective and potent CDK9 inhibitor with an IC50 of 1 nM
Catalog No. 50-002-70191