Learn More
Medchemexpress LLC 4-[[[4-[5-chloro-2-[[4-(2-methoxyethylamino)cyclohexyl]amino]-4-pyridinyl]-1,3-thiazol-2-yl]amino]... | 2247481-21-4 | MFCD31807311 | 98.7% | 505.08 | C24H33ClN6O2S | 1mg
SDP
Supplier:  Medchemexpress LLC HYX01501MG
Encompass_Preferred
JSH-150 is a highly selective and potent CDK9 inhibitor with an IC50 of 1 nM
Catalog No. 50-002-70249