Learn More
Medchemexpress LLC [4-[[4-[5-(cyclopropylmethyl)-1-methyl-pyrazol-4-yl]-5-fluoranyl-pyrimidin-2-yl]amino]cyclohexyl]azanium | 2079069-01-3 | 99.3% | 344.43 | C18H25FN6 | 50MG
SDP
Supplier:  Medchemexpress LLC HY12395550MG
Encompass_Preferred
Casein Kinase inhibitor A86 is a small-molecule research compound that inhibits casein kinase 1α (CK1α) and shows additional activity against CDK7 and CDK9. It has demonstrated induction of leukemia cell apoptosis in preclinical studies and is supplied for laboratory research use only. The compound is provided as a solid and is available ready-made as a DMSO solution for assay use.
Catalog No. 50-002-37386