Learn More
Medchemexpress LLC 4-(4,6-dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride | 3945-69-5 | MFCD03613550 | 99.5% | 276.72 g·mol⁻¹ | C10H17ClN4O3 | 100g
SDP
Supplier:  Medchemexpress LLC HYY1168100G
Encompass_Preferred
DMTMM (4-(4 6-dimethoxy-1 3 5-triazine-2-yl)-4-methylmorpholinium chloride) is a coupling agent DMTMM can activate carboxyl groups and promote the formation of amide bonds DMTMM plays an important role in promoting the chemical modification of biomacromolecules such as polysaccharides and proteins DMTMM can be used for research of tissue engineering breast cancer corneal regeneration and biomaterials[1][2][3][4][5][6][7][8]
Catalog No. 50-002-67047