Learn More
Medchemexpress LLC 4-(4-(6-methoxynaphthalen-2-yl)-2-(4-(methylsulfinyl)phenyl)-1H-imidazol-5-yl)pyridine | 948557-43-5 | MFCD14585804 | >=95.0% | 439.53 g/mol | C26H21N3O2S | 5 MG

Supplier: Medchemexpress LLC HY1005565MG
Tie2 kinase inhibitor 1 is a small-molecule, selective inhibitor of the Tie2 receptor tyrosine kinase with reported anti-cancer activity. It has a reported biochemical IC50 of 250 nM and is supplied for research use.
- Selective Tie2 kinase inhibition with an IC50 of 250 nM.
- Reported anti-cancer activity in preclinical studies.
- Molecular formula C26H21N3O2S and molecular weight 439.53 g/mol.
- CAS number 948557-43-5 for unambiguous identification.
- Purity ≥95% as reported by suppliers.
- Available in small research quantities (example 5 mg).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.