Learn More
Medchemexpress LLC 4-(4-acetyl-phenoxy)-butyric acid | 65623-82-7 | MFCD03658048 | 98.1% | 222.24 | C12H14O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY13226125MG
Encompass_Preferred
AcBut (4-(4-acetyl-phenoxy)-butyric acid) is an acid-labile, cleavable ozogamicin linker used as a building block in antibody-drug conjugate (ADC) synthesis. It provides an acid-sensitive cleavage site for controlled payload release and is supplied as a high-purity solid suitable for conjugation chemistry.
  • Cleavable ozogamicin linker for ADC synthesis.
  • Acid-labile cleavage enables controlled payload release.
  • High purity suitable for conjugation chemistry.
  • Available in 25 mg, 50 mg, and 100 mg packages, larger sizes by quote.
Catalog No. 50-002-03708