Learn More
Medchemexpress LLC 4,4'-Biphthalic anhydride | 2420-87-3 | 98.0% | 294.22 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW008721
Encompass_Preferred
4,4'-Biphthalic anhydride (BPDA) is a rigid symmetrical polyamide. It is used in the production of high-performance polymers, such as polyimide products and their composites.
  • Rigid symmetrical polyamide
  • Used in high-performance polymers
  • Suitable for polyimide products and composites
Catalog No. 50-004-51315