Learn More
Medchemexpress LLC 4,4'-Bis(4-aminophenoxy)biphenyl | 13080-85-8 | 99.9% | 368.44 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW008680
Encompass_Preferred
4,4'-Bis(4-aminophenoxy)biphenyl (Compound 4b) is a phytotoxic biphenyl analogue that inhibits the proliferation of Lemna fronds.
  • Phytotoxic biphenyl analogue
  • Inhibits the proliferation of Lemna fronds
  • Available with high purity
  • Documentation available: Data Sheet, COA, SDS, Handling Instructions, HNMR, RP-HPLC
Catalog No. 50-003-66306