Learn More
Medchemexpress LLC 4,4'-Bis(4-aminophenoxy)biphenyl | 13080-85-8 | 99.9% | 368.44 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW00868050G
Encompass_Preferred
4,4'-Bis(4-aminophenoxy)biphenyl, also referred to as Compound 4b, is a biphenyl analogue. This compound demonstrates phytotoxic characteristics and has been observed to inhibit the proliferation of Lemna fronds.
  • It is a biphenyl analogue.
  • It exhibits phytotoxic properties.
  • It inhibits the proliferation of Lemna fronds.
Catalog No. 50-004-51166