Learn More
Medchemexpress LLC 4-(4-Bromobenzyl)morpholine | 132833-51-3 | 99.9% | C11H14BrNO | 10 G

Supplier: Medchemexpress LLC HY7687710G
4-(4-Bromobenzyl)morpholine is a chemical compound, also known by its CAS Number 132833-51-3. This product is supplied as a white to off-white solid.
- High purity (99.9% by HPLC)
- Chemical formula: C11H14BrNO
- Molecular weight: 256.14
- Solid form for easy handling
- Store at room temperature for 3 years
- Store in solvent at -80°C for 2 years or -20°C for 1 year
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.