Learn More
Medchemexpress LLC 4-(4-Bromobenzyl)morpholine | 132833-51-3 | 99.9% | C11H14BrNO | 50 G
SDP
Supplier:  Medchemexpress LLC HY7687750G
Encompass_Preferred
4-(4-Bromobenzyl)morpholine is a chemical compound with the molecular formula C11H14BrNO and a molecular weight of 256.14. It appears as a white to off-white solid with a high purity of 99.94% as determined by HPLC. This product is suitable for various laboratory applications.
  • Appears as a white to off-white solid
  • Purity of 99.94% (HPLC)
  • Stable for 3 years at room temperature
  • Suitable for laboratory use
Catalog No. 50-004-34017