Learn More
Medchemexpress LLC 4-(4-carbamoylphenoxy)benzamide | 6336-34-1 | 99.8% | 256.26 g/mol | C14H12N2O3 | 100MG

Supplier: Medchemexpress LLC HY123512100MG
OUL35 (NSC39047) is a small-molecule, selective inhibitor of ARTD10 (PARP10) used as a research reagent to probe PARP10 activity and related cellular pathways. It exhibits potent biochemical activity and has characterized solubility and storage properties for use in biochemical and cell-based studies.
- Selective ARTD10 (PARP10) inhibitor with IC50 329 nM.
- Cytoprotective activity in HeLa cells (IC50 1.35 μM).
- High purity (99.8%) and solid, white to off-white appearance.
- Soluble in DMSO (83.33 mg/mL); in vivo formulations ≥ 2.08 mg/mL in 10% DMSO/90% SBE-β-CD or corn oil.
- Store at 4°C and protect from light; in solvent: -80°C (6 months), -20°C (1 month).
- Molecular formula C14H12N2O3; molecular weight 256.26 g/mol.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.