Learn More
Medchemexpress LLC 4',4-Dibromobiphenyl | 92-86-4 | 99.8% | 500 G

Supplier: Medchemexpress LLC HYB1955500G
4',4-Dibromobiphenyl is a biological molecule intended for research use only. It is not sold to patients and must be handled by suitably qualified experienced scientists in appropriately equipped and authorized facilities.
- Purity is 99.82%.
- Molecular weight is 312.00.
- Chemical formula is C12H8Br2.
- CAS number is 92-86-4.
- Appearance is solid.
- Color is white to yellow.
- Store solid at room temperature for 3 years.
- Store in solvent at -80°C for 2 years or -20°C for 1 year.
- HPLC purity is 99.69%.
- Water (KF) is 0.02%.
- Residue on ignition is 0.23%.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.