Learn More
Medchemexpress LLC 4,4-difluorobenzhydrol | 365-24-2 | MFCD00000357 | 98.2% | 220.21 g·mol⁻¹ | C13H10F2O | 100 G
SDP
Supplier:  Medchemexpress LLC HYW037331100G
Encompass_Preferred
4,4-Difluorobenzhydrol is an aromatic secondary alcohol used as a synthetic intermediate in medicinal chemistry. It is employed as a precursor for muscarinic acetylcholine receptor antagonist research and for preparing derivatives that may selectively target the M1 receptor subtype. Supplied as a high-purity solid for laboratory research use.
  • High purity (98.2%) suitable for synthetic chemistry.
  • Solid aromatic alcohol used as a synthetic intermediate.
  • Compatible with common organic transformations in medicinal chemistry.
  • Stable solid form with standard laboratory storage and handling requirements.
  • Molecular formula C13H10F2O and molecular weight 220.21 g·mol⁻¹.
Catalog No. 50-003-01669