Learn More
Medchemexpress LLC 4,4'-dimethoxybenzil | 1226-42-2 | MFCD00008405 | 99.8% | 270.28 g/mol | C16H14O4 | 5 G
SDP
Supplier:  Medchemexpress LLC HY1036105G
Encompass_Preferred
4,4'-Dimethoxybenzil is a high-purity benzil derivative and a potent inhibitor of human intestinal carboxyl esterase (hiCE) with a reported Ki of 70 nM. It is supplied for research use in solid and solution formats and is suitable for biochemical assays, enzyme inhibition studies, and preclinical research.
  • High purity suitable for biochemical and enzyme inhibition studies.
  • Potent hiCE inhibitor (Ki = 70 nM) for target validation and assay development.
  • Available as solid (including 5 g) and as a 10 mM solution in DMSO for convenience.
  • Soluble in DMSO (approx. 50 mg/mL) and practically insoluble in water.
  • Stable when stored as powder at -20°C (up to 3 years) and in solution at -80°C (up to 6 months).
Catalog No. 50-002-47639