Learn More
Medchemexpress LLC 4,4'-dimethoxybenzil (p-anisil) | 1226-42-2 | MFCD00008405 | 99.8% | 270.28 | C16H14O4 | 10 G

Supplier: Medchemexpress LLC HY10361010G
4,4'-Dimethoxybenzil (CAS 1226-42-2) is a small-molecule inhibitor used in biochemical and enzymology research. It is supplied as a high-purity solid and as a DMSO solution for assay use. The compound is intended for research use only and is not for human or clinical applications.
- Acts as a human intestinal carboxyl esterase (hiCE) inhibitor with Ki = 70 nM.
- Molecular formula C16H14O4; molecular weight 270.28.
- High purity (99.83%) suitable for analytical and biochemical assays.
- Appearance: light yellow to green yellow solid.
- Available as a 10 mM solution in DMSO and in solid sizes up to 50 g; bulk quantities available by request.
- Provided for research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.