Learn More
Medchemexpress LLC 4,4-dimethyl-2-cyclopenten-1-one | 22748-16-9 | MFCD00075081 | 99.9% | 110.15 g/mol | C7H10O | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW02460410MM/1ML
Encompass_Preferred
4,4-dimethyl-2-cyclopenten-1-one is a substituted cyclopentenone natural product reported from Apocyniveneti folium and noted in the literature for tumor-selective cytotoxic effects. It is used in biological and chemical research where a small, reactive α,β-unsaturated carbonyl scaffold is required, and is supplied in ready-to-use formats for assays and analytical work.
  • Substituted cyclopentenone small molecule with formula C7H10O.
  • Reported tumor-selective cytotoxicity in cell-based studies.
  • Provided as a 10 mM solution in DMSO for ready-to-use assays.
  • High purity, approximately 99.9% by supplier documentation.
  • Molecular weight 110.15 g/mol.
  • Available as solution and in multiple solid quantities for synthesis or analysis.
Catalog No. 50-003-01619