Learn More
Medchemexpress LLC 4,4-dimethyl-2-cyclopenten-1-one | 22748-16-9 | MFCD00075081 | 99.9% | 110.15 g/mol | C7H10O | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW024604
Encompass_Preferred
4,4-Dimethyl-2-cyclopenten-1-one is an α,β-unsaturated cyclopentenone provided for research applications. It has been reported from plant sources and demonstrated tumor-selective cytotoxicity in cell studies. The product is offered in small laboratory packs and solution formats for biochemical and cell biology experiments.
  • Alpha,beta-unsaturated cyclopentenone structure; electrophilic Michael acceptor properties.
  • Reported tumor-selective cytotoxicity in in vitro cell assays.
  • High purity (>99.9%) suitable for analytical and biological work.
  • Available as small solid packs and as DMSO solution for ready-to-use dosing.
  • Molecular formula C7H10O; molecular weight 110.15 g/mol.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-003-03832