Learn More
Medchemexpress LLC 4-(4-fluoro-2-(1-(4-isopropylphenyl)-5-phenyl-1H-pyrazol-3-yl)phenoxy)butanoic acid | 2988135-14-2 | 99.9% | 458.52 g/mol | C28H27FN2O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1459905MG
Encompass_Preferred
FABPs ligand 6 is a small-molecule inhibitor of fatty acid-binding proteins FABP5 and FABP7 used in preclinical research. It exhibits selective binding affinity (KD ~874 nM for FABP5 and KD ~20 nM for FABP7) and is supplied as a powder or as a DMSO screening solution for laboratory use.
  • Inhibits FABP5 and FABP7 with reported KD values of ~874 nM and ~20 nM.
  • High reported purity, suitable for research applications.
  • Available as powder and as a 10 mM solution in DMSO for screening.
  • Useful for studies of neuroinflammation and multiple sclerosis models.
  • Molecular weight 458.52 g/mol and formula C28H27FN2O3.
Catalog No. 50-002-21182