Learn More
Medchemexpress LLC 4,4'-(methylene-d2)bis(phen-2,3,5,6-d4-ol) | 1794786-93-8 | 99.6% | 210.29 g/mol | C13H2D10O2 | 1 MG

Supplier: Medchemexpress LLC HYW585918S1MG
Deuterium-labeled 4,4'-dihydroxydiphenylmethane supplied as a high-purity research reagent for use as an internal standard or tracer in analytical and mass spectrometry applications. Provided with analytical documentation and recommended storage conditions.
- Deuterium-labeled internal standard suitable for mass spectrometry
- High reported purity of 99.6%
- Chemical formula C13H2D10O2 and molecular weight 210.29 g/mol
- Supplied as a white to off-white solid, available in milligram quantities
- Includes analytical documentation: COA, HPLC, MS, and SDS
- Recommended storage at 4°C under inert atmosphere; in solution store at -80°C for long-term
- Intended for research use only and not for human or clinical applications
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.