Learn More
Medchemexpress LLC 4,4'-Sulfonyldiphenol-d8 | 2483831-28-1 | 99.2% | 258.32 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW011927S1MG
Encompass_Preferred
4,4'-Sulfonyldiphenol-d8 (Bisphenol S (4,4'-Sulfonyldiphenol)-d8) is the deuterium labeled 4,4'-Sulfonyldiphenol. It is a substitute for Bisphenol A, widely used in industrial and consumer products. This compound acts as an estrogen receptor (ER) agonist and can competitively bind to thyroid hormone receptors (TR), affecting breast development and reducing androgen receptor (AR) expression in fetal testes. It promotes glioblastoma progression by upregulating the EZH2 mediated PI3K/AKT/mTOR pathway. Chronic exposure can lead to lipid deposition and dyslipidemia in the mouse liver, contributing to obesity. It also induces intestinal inflammation and accelerates atherosclerosis progression.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-53956