Learn More
Medchemexpress LLC 4-[5-(4-chlorophenyl)-2-methyl-3-propanoylpyrrol-1-yl]benzenesulfonamide | 1000279-69-5 | 99.8% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1214910MM 1ML
Encompass_Preferred
A-867744 is a research chemical that acts as a type II positive allosteric modulator of the alpha7 nicotinic acetylcholine receptor (α7 nAChR), enhancing agonist-evoked responses with reported EC50 ≈ 1.0 μM. It is supplied as a high-purity research-grade compound and is commonly provided as a 10 mM solution in DMSO for biochemical assays.
  • Type II positive allosteric modulator of α7 nAChR.
  • Reported EC50 ≈ 1.0 μM in FLIPR calcium assays.
  • Chemical formula C20H19ClN2O3S and molecular weight 402.89 g/mol.
  • High purity (>99.7%) suitable for research use.
  • Available as a 10 mM solution in DMSO and as solid quantities for formulation.
Catalog No. 50-002-32512