Learn More
Medchemexpress LLC 4-[(5,5-dimethyl-8-quinolin-3-yl-6H-naphthalene-2-carbonyl)amino]benzoic acid | 182135-66-6 | MFCD00952209 | 99.5% | 448.5 g/mol | C29H24N2O3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1012591MG
Encompass_Preferred
This compound is a small-molecule neutral antagonist selective for retinoic acid receptor alpha (RARα) used in research to modulate retinoid signaling. It exhibits high affinity for RARα and has reported effects on inflammatory signaling, angiogenesis, cell migration, and blue-light-induced phototoxicity.
  • High affinity for RARα (Ki ≈ 2.5 nM).
  • High purity (>99%).
  • Molecular formula C29H24N2O3 and molecular weight 448.5 g/mol.
  • Suitable for in vitro and in vivo studies of inflammation, angiogenesis, and cell migration.
  • Reported to reduce blue-light-induced phototoxicity.
Catalog No. 50-002-12494