Learn More
Medchemexpress LLC 4-[(5,5-dimethyl-8-quinolin-3-yl-6H-naphthalene-2-carbonyl)amino]benzoic acid | 182135-66-6 | MFCD00952209 | 99.5% | 448.50 g/mol | C29H24N2O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10125950MG
Encompass_Preferred
A neutral, RARα-selective antagonist used as a research reagent to modulate retinoic acid receptor alpha signaling in biochemical and cell-based studies. It is supplied as a high-purity powder suitable for analytical and biological assays, with reported activity in inflammatory and angiogenic pathways and effects on cell migration and phototoxicity.
  • High affinity for RARα receptor, enabling receptor-specific studies
  • High purity suitable for analytical and biological assays
  • Powder form allows flexible dosing and formulation
  • Demonstrated modulation of inflammatory signaling pathways
  • Applicable to studies of phototoxicity, angiogenesis, and cell migration
Catalog No. 50-002-12945