Learn More
Medchemexpress LLC 4,5,6,7-tetrafluoro-L-tryptophan | 72120-72-0 | 276.19 g·mol⁻¹ | C11H8F4N2O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY17174610MG
Encompass_Preferred
F4-Trp (4,5,6,7-tetrafluoro-L-tryptophan) is a synthetic fluorinated amino acid used as a research ligand for the α3β4 nicotinic acetylcholine receptor. It is employed in studies of neurotransmission, addiction, and related neurological processes, and is provided as a solid powder with recommended sealed, low-temperature storage for stability.
  • Ligand for α3β4 nicotinic acetylcholine receptor.
  • Useful for neurotransmission and addiction research.
  • Provided as a solid powder for consistent handling.
  • Stable when stored sealed at low temperatures.
  • Available in small mg-scale quantities suitable for screening and assay work.
Catalog No. 50-002-95162