Learn More
Medchemexpress LLC 4 5 -Dibromofluores 10mM 1mL | 596-03-2 | 1 ML

Supplier: Medchemexpress LLC HY11490710MM 1ML
4',5'-Dibromofluorescein is a brominated fluorescein derivative used as a fluorescent dye and as a ligand for studying protein properties by spectral analysis. It is supplied as a solid powder and as a ready-to-use 10 mM solution in DMSO for research applications.
- Fluorescent xanthene dye with absorption and emission properties suitable for microscopy and spectral assays.
- CAS 596-03-2; molecular formula C20H10Br2O5; molecular weight 490.10 g·mol⁻¹.
- Dye content/purity typically 95.0% (dye basis).
- Available as a 10 mM solution in DMSO (1 mL) and in solid form in multiple mass quantities.
- Used as a ligand in protein spectral studies, as a staining reagent, and as a fluorescence tracer in analytical applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.