Learn More
Apexbio Technology LLC 4,5-Dicaffeoylquinic acid 57378-72-0 5mg
SDP
Supplier:  Apexbio Technology LLC B84585
Encompass_Preferred
4 5-Dicaffeoylquinic acid (CAS 57378-72-0) is a small-molecule inhibitor targeting -glucosidase It is designed to inhibit -glucosidase enzyme activity thereby modulating glucose metabolism 4 5-Dicaffeoylquinic acid exerts its biological activity primarily through enzyme inhibition In vitro studies demonstrate -glucosidase inhibition with an IC50 value of approximately 20 M In cell-based assays it also mediates antiproliferative effects via cell cycle arrest in prostate cancer cell lines Based on these pharmacological properties 4 5-Dicaffeoylquinic acid holds research potential in diabetes cancer proliferation and antiviral therapeutic investigations
Catalog No. 50-000-91174