Learn More
Medchemexpress LLC 4',5-Dihydroxyflavone | 6665-67-4 | 254.24 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN1881
Encompass_Preferred
4',5-Dihydroxyflavone (Compound 2c; Compound B3) is a flavonoid compound. It can inhibit various oxidases, with IC50 values of less than 1 μM for collagenase A (ColA) and 66 μM for α-glucosidase. Its Ki value for soybean lipoxygenase-1 (LOX-1) is 102.6 μM. This compound is suitable for studies on anti-toxicity and anti-diabetes. 4',5-Dihydroxyflavone is also a potent inhibitor of ColA, a bacterial virulence factor that aids bacterial spread by degrading host collagen proteins.
  • Type of flavonoid compound.
  • Inhibits various oxidases.
  • IC50 values for collagenase A (ColA) and α-glucosidase are < 1 μM and 66 μM, respectively.
  • Ki value for soybean lipoxygenase-1 (LOX-1) is 102.6 μM.
  • Can be used in studies on anti-toxicity and anti-diabetes.
  • Potent inhibitor of ColA, a bacterial virulence factor.
Catalog No. 50-003-90001