Learn More
Cayman Chemical 4 5-EpDPA methyl ester 100UG
SDP
Supplier:  Cayman Chemical 90314100 UG
Encompass_Preferred
A stable derivative of (±)4(5)-EpDPA, a CYP450 metabolite of DHA which can be further metabolized to the diol metabolite; the active free acid can be regenerated from the methyl ester by careful base hydrolysis
Catalog No. 50-289-4963