Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Permegear 4.5L HEATER/CIRCULATOR 115V
SDP

Supplier:  Permegear 1006001

Encompass

  • 4.5 L reservoir volume.
  • Designed for temperature control for diffusion cell systems.
  • Maintains stable temperature between 20ºC to 150ºC.
  • Exceptional temperature stability of ±0.03ºC.
  • Flow rate of 15 L/min.
  • Operates at a power input of 115V.
  • Integrates with PermeGear automated systems.

Catalog No. NC3038674


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.