Learn More
S2 Media 4.5mL Tryptic Soy Agar (TSA)/Soybean-Casein Digest (SCD) Slants in 16x115mm Polypropylene Flat Bottom Tube, 50/box
Supplier:  S2 Media 5246
Encompass_Preferred
Tryptic Soy Agar (TSA) is a non-selective and non-differential, general growth medium. It is recommended for the isolation and cultivation of fastidious and non-fastidious microorganisms, although it is not the medium of choice for anaerobes. TSA is also recommended for the preparation, maintenance, and storage of test strains. • Product complies with AOAC, BAM, CLSI, ISO, Harmonized USP/EP/JP, SMWW, and USDA reference methods and performance criteria.• TSA should be used by trained professionals within a laboratory setting.• For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5754