Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-[6-(1-Azetidinyl)-1,2-dihydro-2-methyl-1-oxo-2,7-naphthyridin-4-yl]-2,6-dimethoxybenzaldeh... | 2460248-49-9 | 97.5% | C21H21N3O4 | 250 MG
SDP

Supplier:  Medchemexpress LLC 250MGHY49393

Encompass_Preferred

BRD9 ligand-6 is a PROTAC target protein ligand that can be utilized for the synthesis of FHD-609.

  • PROTAC target protein ligand
  • Suitable for FHD-609 synthesis
  • Molecular weight: 379.41
  • Appearance: Off-white to gray solid
  • Purity: 97.5%
  • Storage: Powder at -20°C for 3 years or 4°C for 2 years; in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-58448


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.