Learn More
Medchemexpress LLC 4-[6-[4-(1-methylethoxy)phenyl]pyrazolo[1,5-a]pyrimidin-3-yl]-quinoline | 1206711-16-1 | 99.8% | 5 MG
SDP
Supplier:  Medchemexpress LLC HY12273R5MG
Encompass_Preferred
DMH-1 (Standard) is an analytical standard of DMH-1, a selective bone morphogenetic protein (BMP) type I receptor inhibitor used as a reference in biochemical and cell-based assays. It exhibits IC50s of 27 nM (ALK1), 107.9 nM (ALK2), <5 nM (ALK3), and 47.6 nM (ALK6).
  • Analytical standard for assay calibration and reference.
  • Selective BMP type I receptor inhibitor targeting ALK1, ALK2, ALK3, and ALK6.
  • Documented IC50s for ALK receptors support target-specific studies.
  • High-purity solid suitable for analytical use (99.78% by supplier certificate).
  • Available in small milligram quantities for research applications.
Catalog No. 50-002-30139