Learn More
Medchemexpress LLC 4-[6-[[4-(cyclopropylmethyl)piperazin-1-yl]methyl]-2-(5-fluoro-1H-indol-4-yl)thieno[3,2-d]pyrimidin | 955977-50-1 | 99.0% | 25 MG

Supplier: Medchemexpress LLC HY1223525MG
PI-3065 is a selective small-molecule inhibitor of PI3K p110δ used as a research tool for biochemical and cellular studies. It displays low-nanomolar potency against p110δ and substantially reduced activity against other class I PI3K isoforms.
- Selective p110δ inhibitor (IC50 = 5 nM; Ki = 1.5 nM).
- Reduced activity against p110α (IC50 910 nM), p110β (IC50 600 nM), and p110γ (IC50 >10,000 nM).
- Molecular formula C27H31FN6OS and molecular weight 506.64 g/mol.
- High reported purity (~98.97%).
- Soluble in DMSO (25 mg/mL; 49.34 mM).
- Store powder at -20°C for long-term storage or 4°C for short-term storage.
- Intended for research use in biochemical assays and cell-based PI3Kδ signaling studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.