Learn More
Medchemexpress LLC 4-(6-(4-(piperazin-1-yl)phenyl)pyrazolo[1,5-a]pyrimidin-3-yl)quinoline hydrochloride | 1062368-62-0 | >98.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY12071C100MG
Encompass_Preferred
LDN193189 hydrochloride is the monohydrochloride salt of LDN-193189, a selective inhibitor of bone morphogenetic protein (BMP) type I receptors used for in vitro research. It is supplied as a solid biochemical reagent for pathway and cell-based assays and is provided with high purity suitable for research use only.
  • Selective inhibitor of BMP type I receptors (ALK1, ALK2, ALK3, ALK6)
  • High purity, ≥98.0% by HPLC
  • Solid form suitable for dissolution in DMSO for in vitro assays
  • Molecular weight 442.9 g/mol; formula C25H23ClN6
  • Commonly used in cell-based and biochemical BMP pathway studies
Catalog No. 50-002-34833