Learn More
Medchemexpress LLC 4-[6-(4-piperazin-1-ylphenyl)pyrazolo[1,5-a]pyrimidin-3-yl]quinoline | 1062368-24-4 | >98.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY12071G50MG
Encompass_Preferred
LDN193189 (GMP) is a GMP-grade small-molecule inhibitor of BMP type I receptors, supplied as a research reagent intended for use as an auxiliary reagent in cell therapy manufacture and regulated research. The compound has molecular formula C25H22N6 and molecular weight 406.49 g/mol, and is provided in a 50 MG package.
  • GMP-grade formulation suitable as an auxiliary reagent in cell therapy manufacture.
  • Selective BMP type I receptor inhibitor for research applications.
  • CAS 1062368-24-4 and molecular formula C25H22N6.
  • Provided as a 50 MG package for controlled dosing and handling.
  • Analytical purity greater than 98.0% by HPLC (supplier reported).
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-30396