Learn More
Strem, An Ascensus Company CAS# 261733-18-0. 2g. 4,6-Bis(diphenylphosphino)phenoxazine, min. 98% NIXANTPHOS. MFCD03788937
SDP
Supplier:  Strem, An Ascensus Company 1504372G
Encompass
CAS# 261733-18-0. 2g. 4,6-Bis(diphenylphosphino)phenoxazine, min. 98% NIXANTPHOS. MFCD03788937. Molecular Weight: 551.55. Molecular Formula: C36H27NOP2. Color/form: white to off-white pwdr. Strem# 15-0437. http://www.strem.com/catalog/v/15-0437/
Catalog No. 50-901-09872