Learn More
Medchemexpress LLC 4,6-Dichloro-N-methylpyridazine-3-carboxamide-d3 | 1609393-89-6 | 98.17% | C6H2D3Cl2N3O | 10 G
SDP
Supplier:  Medchemexpress LLC HY45842S10G
Encompass_Preferred
4,6-Dichloro-N-methylpyridazine-3-carboxamide-d3 is a deuterium labeled compound for research use only. This yellow to brown solid functions as a tracer and an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS. It is recommended for storage at -20°C in a sealed container, away from moisture, with specific solvent storage guidelines for -80°C (6 months) or -20°C (1 month).
  • Deuterium labeled for research applications
  • Useful as a tracer in various analyses
  • Serves as an internal standard for NMR, GC-MS, and LC-MS
  • Presented as a yellow to brown solid
  • Optimal storage at -20°C, sealed, and away from moisture
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month
  • Requires handling with protective gloves, clothing, and eye protection
  • Use in well-ventilated areas to avoid inhalation
Catalog No. 50-003-59167