Learn More
Medchemexpress LLC 4,6-Dichloronicotinic acid | 73027-79-9 | 192.00 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00255310G
Encompass_Preferred
4,6-Dichloronicotinic acid is a biochemical reagent presented as a white to light yellow powder to crystal, recommended for storage at room temperature, below 15°C in a cool and dark place. It serves as a biological material or organic compound for life science-related research.
  • Biochemical reagent
  • White to light yellow powder to crystal
  • Purity of >98.0%
  • Suitable for life science research
Catalog No. 50-004-45901